Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604692 MIM: 609475 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKAP8 - A-kinase anchoring protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291478.1 | 1654 | Intron | NP_001278407.1 | |||
NM_014371.3 | 1654 | Intron | NP_055186.3 | |||
XM_005259854.3 | 1654 | Intron | XP_005259911.2 | |||
XM_011527916.2 | 1654 | Missense Mutation | ACA,GCA | T,A 517 | XP_011526218.1 |
Set Membership: |
HapMap |