Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTGTATTGAGGTTTCAGGTGA[A/C]CAGGCCATATTGAAACTTGATCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300435 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985692 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC107985692 - A-kinase anchor protein 17B-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGRMC1 - progesterone receptor membrane component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282621.1 | Intron | NP_001269550.1 | ||||
NM_006667.4 | Intron | NP_006658.1 |
Set Membership: |
HapMap Validated |