Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610002 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL21A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL21A1 - collagen type XXI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318751.1 | 3755 | UTR 3 | NP_001305680.1 | |||
NM_001318752.1 | 3755 | UTR 3 | NP_001305681.1 | |||
NM_001318753.1 | 3755 | UTR 3 | NP_001305682.1 | |||
NM_001318754.1 | 3755 | UTR 3 | NP_001305683.1 | |||
NM_030820.3 | 3755 | UTR 3 | NP_110447.2 | |||
XM_006715223.1 | 3755 | UTR 3 | XP_006715286.1 | |||
XM_011514924.2 | 3755 | UTR 3 | XP_011513226.1 | |||
XM_011514925.2 | 3755 | UTR 3 | XP_011513227.1 | |||
XM_011514926.1 | 3755 | UTR 3 | XP_011513228.1 | |||
XM_011514927.1 | 3755 | UTR 3 | XP_011513229.1 |
Set Membership: |
HapMap |