Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTGCTGCTGTAACAAGAATAAA[C/T]AGTATTGAAATGTCTAGAGCACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608618 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM3C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM3C - family with sequence similarity 3 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040020.1 | Intron | NP_001035109.1 | ||||
NM_014888.2 | Intron | NP_055703.1 | ||||
XM_011515736.2 | Intron | XP_011514038.1 | ||||
XM_011515737.2 | Intron | XP_011514039.1 |
Set Membership: |
Validated |