Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTGGTCAGTGGCCTGGACTAG[C/T]CTGGGAACTGTTGGAACATGGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614560 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAU2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAU2 - MAU2 sister chromatid cohesion factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015329.3 | Intron | NP_056144.3 | ||||
XM_005259837.3 | Intron | XP_005259894.2 | ||||
XM_005259838.3 | Intron | XP_005259895.3 | ||||
XM_006722711.3 | Intron | XP_006722774.2 | ||||
XM_011527844.2 | Intron | XP_011526146.1 | ||||
XM_017026539.1 | Intron | XP_016882028.1 |