Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGACTCTTGTTCCAGCTCTCC[A/G]CCCTGCTTGCTGGCAGGCAGGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300019 MIM: 300059 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HCFC1 - host cell factor C1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCFC1-AS1 - HCFC1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3202-1 - microRNA 3202-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3202-2 - microRNA 3202-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM187 - transmembrane protein 187 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003492.2 | Intron | NP_003483.1 | ||||
XM_011531199.2 | Intron | XP_011529501.1 |