Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGTGCTCTGCCCTTTTTTCCC[C/T]CCGCTTCCTTCTGCAGCGGGGTCGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602386 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF212 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF212 - zinc finger protein 212 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012256.3 | 318 | Intron | NP_036388.2 | |||
XM_005250055.2 | 318 | Intron | XP_005250112.2 | |||
XM_011516581.2 | 318 | Intron | XP_011514883.2 | |||
XM_011516582.1 | 318 | Intron | XP_011514884.1 | |||
XM_017012646.1 | 318 | Intron | XP_016868135.1 | |||
XM_017012647.1 | 318 | Intron | XP_016868136.1 | |||
XM_017012648.1 | 318 | UTR 5 | XP_016868137.1 |
Set Membership: |
HapMap |