Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGTTCCACAGTTTGGCTTATC[A/G]GTTACTGTAGCCACAGACGTCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609246 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927051 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101927051 - uncharacterized LOC101927051 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDXP - pyridoxal phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020315.4 | Intron | NP_064711.1 |
SH3BP1 - SH3 domain binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |