Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607880 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC6B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXOC6B - exocyst complex component 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321729.1 | Intron | NP_001308658.1 | ||||
NM_001321730.1 | Intron | NP_001308659.1 | ||||
NM_001321731.1 | Intron | NP_001308660.1 | ||||
NM_001321733.1 | Intron | NP_001308662.1 | ||||
NM_001321734.1 | Intron | NP_001308663.1 | ||||
NM_015189.2 | Intron | NP_056004.1 | ||||
XM_005264224.1 | Intron | XP_005264281.1 | ||||
XM_011532711.2 | Intron | XP_011531013.1 | ||||
XM_011532712.2 | Intron | XP_011531014.1 | ||||
XM_017003641.1 | Intron | XP_016859130.1 | ||||
XM_017003642.1 | Intron | XP_016859131.1 |
Set Membership: |
HapMap |