Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601951 MIM: 615811 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CLK1 - CDC like kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL3 - peptidylprolyl isomerase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032472.3 | 701 | Missense Mutation | GAG,GAT | E,D 150 | NP_115861.1 | |
NM_130906.2 | 701 | Missense Mutation | GAG,GAT | E,D 146 | NP_570981.1 | |
XM_005246651.3 | 701 | Missense Mutation | GAG,GAT | E,D 150 | XP_005246708.1 | |
XM_005246652.4 | 701 | Missense Mutation | GAG,GAT | E,D 146 | XP_005246709.1 | |
XM_011511358.2 | 701 | Missense Mutation | GAG,GAT | E,D 146 | XP_011509660.1 | |
XM_017004352.1 | 701 | Missense Mutation | GAG,GAT | E,D 150 | XP_016859841.1 | |
XM_017004353.1 | 701 | Missense Mutation | GAG,GAT | E,D 150 | XP_016859842.1 | |
XM_017004354.1 | 701 | Intron | XP_016859843.1 | |||
XM_017004355.1 | 701 | Intron | XP_016859844.1 | |||
XM_017004356.1 | 701 | Intron | XP_016859845.1 | |||
XM_017004357.1 | 701 | Missense Mutation | GAG,GAT | E,D 97 | XP_016859846.1 | |
XM_017004358.1 | 701 | Missense Mutation | GAG,GAT | E,D 97 | XP_016859847.1 | |
XM_017004359.1 | 701 | Missense Mutation | GAG,GAT | E,D 97 | XP_016859848.1 | |
XM_017004360.1 | 701 | Missense Mutation | GAG,GAT | E,D 97 | XP_016859849.1 |
Set Membership: |
HapMap Validated |