Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602245 MIM: 615323 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTPBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GTPBP1 - GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004286.4 | 1051 | Intron | NP_004277.2 | |||
XM_011530537.2 | 1051 | UTR 5 | XP_011528839.1 | |||
XM_017029099.1 | 1051 | UTR 5 | XP_016884588.1 | |||
XM_017029100.1 | 1051 | UTR 5 | XP_016884589.1 | |||
XM_017029101.1 | 1051 | Intron | XP_016884590.1 | |||
XM_017029102.1 | 1051 | Intron | XP_016884591.1 | |||
XM_017029103.1 | 1051 | Intron | XP_016884592.1 |
JOSD1 - Josephin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |