Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGGGGAGTGCAGGATCTGCCT[C/T]GCACCTCTGCCTACTGCAGTGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188070 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACTIN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CACTIN - cactin, spliceosome C complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080543.1 | Intron | NP_001074012.1 | ||||
NM_021231.1 | Intron | NP_067054.1 | ||||
XM_011528160.2 | Intron | XP_011526462.1 | ||||
XM_011528161.2 | Intron | XP_011526463.1 |
CACTIN-AS1 - CACTIN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXA2R - thromboxane A2 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |