Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCTGTAAGAGTTCTCTTAGCG[G/T]TCCAAGAACCTGTGTCTAATTACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602740 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370027 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105370027 - uncharacterized LOC105370027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAB1 - protein kinase AMP-activated non-catalytic subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006253.4 | Intron | NP_006244.2 | ||||
XM_005253909.1 | Intron | XP_005253966.1 |
Set Membership: |
HapMap |