Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGGCACGATCTCGGCTCACTG[C/T]AAGCAATTGTCCCGCCTCAGCCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147574 MIM: 608193 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IPO4 - importin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRF9 - interferon regulatory factor 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REC8 - REC8 meiotic recombination protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048205.1 | Intron | NP_001041670.1 | ||||
NM_005132.2 | Intron | NP_005123.2 | ||||
XM_011537421.1 | Intron | XP_011535723.1 | ||||
XM_017021841.1 | Intron | XP_016877330.1 |