Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGTTTAGGAGTTAGCATTAGG[A/G]GAGAGGTGTCTGTAAGGAACTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606276 MIM: 614189 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRTAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRTAC1 - cartilage acidic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GOLGA7B - golgin A7 family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010917.2 | Intron | NP_001010917.1 |
LINC00866 - long intergenic non-protein coding RNA 866 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |