Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACCCTTCCACAATCATAATCTTC[A/G]CCCTCTTTGGAGCTAATGGTTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUT10 - fucosyltransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032664.3 | Intron | NP_116053.3 | ||||
XM_011544676.2 | Intron | XP_011542978.1 | ||||
XM_011544677.2 | Intron | XP_011542979.1 | ||||
XM_011544678.2 | Intron | XP_011542980.1 | ||||
XM_011544679.2 | Intron | XP_011542981.1 | ||||
XM_011544680.2 | Intron | XP_011542982.1 | ||||
XM_011544681.2 | Intron | XP_011542983.1 |