Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCAAAATGAATTTGTGTCCTTG[C/T]TCATCTAATTTTCCTAGTAAATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616931 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FUT10 - fucosyltransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032664.3 | Intron | NP_116053.3 | ||||
XM_011544676.2 | Intron | XP_011542978.1 | ||||
XM_011544677.2 | Intron | XP_011542979.1 | ||||
XM_011544678.2 | Intron | XP_011542980.1 | ||||
XM_011544679.2 | Intron | XP_011542981.1 | ||||
XM_011544680.2 | Intron | XP_011542982.1 | ||||
XM_011544681.2 | Intron | XP_011542983.1 |
Set Membership: |
HapMap |