Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGTTTAATACGGGTTTAGAAAC[A/G]AAAATGAGAATTCCACGCAATAAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190070 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRAS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KRAS - KRAS proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004985.4 | Intron | NP_004976.2 | ||||
NM_033360.3 | Intron | NP_203524.1 | ||||
XM_006719069.3 | Intron | XP_006719132.1 | ||||
XM_011520653.2 | Intron | XP_011518955.1 | ||||
XM_017019292.1 | Intron | XP_016874781.1 | ||||
XM_017019293.1 | Intron | XP_016874782.1 |
LYRM5 - LYR motif containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |