Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190070 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRAS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KRAS - KRAS proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004985.4 | 1883 | UTR 3 | NP_004976.2 | |||
NM_033360.3 | 1883 | UTR 3 | NP_203524.1 | |||
XM_006719069.3 | 1883 | Intron | XP_006719132.1 | |||
XM_011520653.2 | 1883 | Intron | XP_011518955.1 | |||
XM_017019292.1 | 1883 | Intron | XP_016874781.1 | |||
XM_017019293.1 | 1883 | UTR 3 | XP_016874782.1 |
LYRM5 - LYR motif containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |