Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCCCTCAAACTTAATATGTTT[C/G]TTAAACAAATTGAAGCCACTAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603217 | ||||||||||||||||||||
Literature Links: |
STX7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STX7 - syntaxin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326578.1 | 3016 | UTR 3 | NP_001313507.1 | |||
NM_001326579.1 | 3016 | UTR 3 | NP_001313508.1 | |||
NM_001326580.1 | 3016 | Intron | NP_001313509.1 | |||
NM_003569.2 | 3016 | UTR 3 | NP_003560.2 | |||
XM_011536178.1 | 3016 | Intron | XP_011534480.1 |