Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCTTGGTTATGAGAAGGAAAG[C/T]GAAAATCAATTTTGACCTGTCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP10D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATP10D - ATPase phospholipid transporting 10D (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020453.3 | Intron | NP_065186.3 | ||||
XM_005248119.4 | Intron | XP_005248176.1 | ||||
XM_005248120.4 | Intron | XP_005248177.1 | ||||
XM_011513722.2 | Intron | XP_011512024.1 | ||||
XM_017008472.1 | Intron | XP_016863961.1 |
Set Membership: |
HapMap Validated |