Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACAATCTTAGAACTCTTGGAGT[A/G]GGGGTAGGAGCTTGCATTGTAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNKS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNKS2 - tankyrase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025235.3 | Intron | NP_079511.1 | ||||
XM_005270185.4 | Intron | XP_005270242.2 | ||||
XM_011540213.1 | Intron | XP_011538515.1 | ||||
XM_011540215.2 | Intron | XP_011538517.1 | ||||
XM_017016696.1 | Intron | XP_016872185.1 | ||||
XM_017016697.1 | Intron | XP_016872186.1 | ||||
XM_017016698.1 | Intron | XP_016872187.1 | ||||
XM_017016699.1 | Intron | XP_016872188.1 | ||||
XM_017016700.1 | Intron | XP_016872189.1 | ||||
XM_017016701.1 | Intron | XP_016872190.1 |
TNKS2-AS1 - TNKS2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |