Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTAAGTTGTTTTATCTGGCAGC[A/G]GGATTTTAAATTTTGTATTCATTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BTNL9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTNL9 - butyrophilin like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308245.1 | Intron | NP_001295174.1 | ||||
NM_152547.4 | Intron | NP_689760.2 | ||||
XM_006714823.3 | Intron | XP_006714886.1 | ||||
XM_006714824.3 | Intron | XP_006714887.1 | ||||
XM_006714825.3 | Intron | XP_006714888.1 | ||||
XM_011534444.2 | Intron | XP_011532746.1 | ||||
XM_017009113.1 | Intron | XP_016864602.1 | ||||
XM_017009114.1 | Intron | XP_016864603.1 | ||||
XM_017009115.1 | Intron | XP_016864604.1 | ||||
XM_017009116.1 | Intron | XP_016864605.1 | ||||
XM_017009117.1 | Intron | XP_016864606.1 |
MIR8089 - microRNA 8089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |