Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614720 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDK19 - cyclin dependent kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300960.1 | 4978 | UTR 3 | NP_001287889.1 | |||
NM_001300963.1 | 4978 | UTR 3 | NP_001287892.1 | |||
NM_001300964.1 | 4978 | UTR 3 | NP_001287893.1 | |||
NM_015076.4 | 4978 | UTR 3 | NP_055891.1 | |||
XM_005266871.3 | 4978 | UTR 3 | XP_005266928.1 | |||
XM_011535630.2 | 4978 | UTR 3 | XP_011533932.1 | |||
XM_011535631.2 | 4978 | UTR 3 | XP_011533933.1 | |||
XM_011535632.1 | 4978 | Intron | XP_011533934.1 | |||
XM_017010587.1 | 4978 | UTR 3 | XP_016866076.1 | |||
XM_017010588.1 | 4978 | UTR 3 | XP_016866077.1 |
Set Membership: |
HapMap Validated |