Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACAGTTTTAACAATGATCTCTT[A/G]CAACATGGAAAGGGACTTACTTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD12B - abhydrolase domain containing 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206673.1 | Intron | NP_001193602.1 | ||||
NM_181533.3 | Intron | NP_853511.2 | ||||
NM_181814.1 | Intron | NP_861535.1 | ||||
XM_006720052.1 | Intron | XP_006720115.1 | ||||
XM_011536472.1 | Intron | XP_011534774.1 | ||||
XM_011536474.2 | Intron | XP_011534776.1 | ||||
XM_017021031.1 | Intron | XP_016876520.1 |
Set Membership: |
HapMap |