Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613134 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TSPAN3 - tetraspanin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168412.1 | 877 | Silent Mutation | GTA,GTT | V,V 137 | NP_001161884.1 | |
NM_005724.5 | 877 | Silent Mutation | GTA,GTT | V,V 201 | NP_005715.1 | |
NM_198902.2 | 877 | Silent Mutation | GTA,GTT | V,V 176 | NP_944492.1 | |
XM_017021857.1 | 877 | Silent Mutation | GTA,GTT | V,V 162 | XP_016877346.1 |