Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATCTTAATCACGATCCCATCGTG[C/T]GCTGAGAGAGGGTGGGCAGGGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF777 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF777 - zinc finger protein 777 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015694.2 | 1262 | Silent Mutation | GCA,GCG | A,A 363 | NP_056509.2 | |
XM_005249980.3 | 1262 | Silent Mutation | GCA,GCG | A,A 407 | XP_005250037.1 | |
XM_011516055.2 | 1262 | Silent Mutation | GCA,GCG | A,A 363 | XP_011514357.1 |
Set Membership: |
HapMap Validated |