Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603423 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PRDM1 - PR domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198.3 | Intron | NP_001189.2 | ||||
NM_182907.2 | Intron | NP_878911.1 | ||||
XM_006715550.3 | Intron | XP_006715613.1 | ||||
XM_011536062.2 | Intron | XP_011534364.1 | ||||
XM_011536063.2 | Intron | XP_011534365.1 | ||||
XM_011536064.2 | Intron | XP_011534366.1 | ||||
XM_017011187.1 | Intron | XP_016866676.1 |
Set Membership: |
HapMap Validated |