Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGAAAAGATTAGTTTTGGGAGC[C/T]GACACAAGAATTCAGTTTGTGCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 248611 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCKDHB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BCKDHB - branched chain keto acid dehydrogenase E1 subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000056.4 | Intron | NP_000047.1 | ||||
NM_001318975.1 | Intron | NP_001305904.1 | ||||
NM_183050.3 | Intron | NP_898871.1 | ||||
XM_005248756.4 | Intron | XP_005248813.1 | ||||
XM_011536023.2 | Intron | XP_011534325.1 | ||||
XM_011536024.2 | Intron | XP_011534326.1 | ||||
XM_011536025.2 | Intron | XP_011534327.1 | ||||
XM_011536026.2 | Intron | XP_011534328.1 |
Set Membership: |
HapMap |