Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGTGTTTGTGCAGTGACCTAA[C/T]ACAGAGAACACAGAGTTGAAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610986 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927751 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101927751 - uncharacterized LOC101927751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRK1 - leucine rich repeat kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024652.4 | Intron | NP_078928.3 | ||||
XM_005254979.3 | Intron | XP_005255036.1 | ||||
XM_011522012.2 | Intron | XP_011520314.1 | ||||
XM_011522013.2 | Intron | XP_011520315.1 | ||||
XM_011522014.2 | Intron | XP_011520316.1 | ||||
XM_011522015.2 | Intron | XP_011520317.1 | ||||
XM_011522016.2 | Intron | XP_011520318.1 | ||||
XM_011522017.2 | Intron | XP_011520319.1 | ||||
XM_011522018.2 | Intron | XP_011520320.1 | ||||
XM_011522019.2 | Intron | XP_011520321.1 | ||||
XM_017022570.1 | Intron | XP_016878059.1 |
Set Membership: |
HapMap |