Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606696 MIM: 603473 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KATNA1 - katanin catalytic subunit A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270519.1 | 7251 | Intron | NP_001257448.1 | |||
NM_004690.3 | 7251 | UTR 3 | NP_004681.1 | |||
XM_006715603.3 | 7251 | Intron | XP_006715666.1 | |||
XM_011536252.2 | 7251 | Intron | XP_011534554.1 | |||
XM_017011474.1 | 7251 | UTR 3 | XP_016866963.1 | |||
XM_017011475.1 | 7251 | UTR 3 | XP_016866964.1 | |||
XM_017011476.1 | 7251 | UTR 3 | XP_016866965.1 | |||
XM_017011477.1 | 7251 | Intron | XP_016866966.1 | |||
XM_017011478.1 | 7251 | UTR 3 | XP_016866967.1 | |||
XM_017011479.1 | 7251 | Intron | XP_016866968.1 | |||
XM_017011480.1 | 7251 | UTR 3 | XP_016866969.1 | |||
XM_017011481.1 | 7251 | Intron | XP_016866970.1 | |||
XM_017011482.1 | 7251 | Intron | XP_016866971.1 |
Set Membership: |
HapMap Validated |