Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609785 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFCP2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFCP2L1 - transcription factor CP2-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014553.2 | 2292 | UTR 3 | NP_055368.1 | |||
XM_017003902.1 | 2292 | UTR 3 | XP_016859391.1 | |||
XM_017003903.1 | 2292 | UTR 3 | XP_016859392.1 | |||
XM_017003904.1 | 2292 | Intron | XP_016859393.1 | |||
XM_017003905.1 | 2292 | Intron | XP_016859394.1 |
Set Membership: |
Validated |