Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612768 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FNIP2 - folliculin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323916.1 | Intron | NP_001310845.1 | ||||
NM_020840.2 | Intron | NP_065891.1 | ||||
XM_005263156.1 | Intron | XP_005263213.1 | ||||
XM_005263158.1 | Intron | XP_005263215.1 | ||||
XM_005263160.3 | Intron | XP_005263217.1 | ||||
XM_011532154.2 | Intron | XP_011530456.1 | ||||
XM_017008487.1 | Intron | XP_016863976.1 |
Set Membership: |
HapMap |