Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613384 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNL1 - cyclin L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308185.1 | Intron | NP_001295114.1 | ||||
NM_020307.3 | Intron | NP_064703.1 | ||||
XM_005247648.3 | Intron | XP_005247705.1 | ||||
XM_006713710.3 | Intron | XP_006713773.1 | ||||
XM_011513011.2 | Intron | XP_011511313.1 | ||||
XM_017006891.1 | Intron | XP_016862380.1 | ||||
XM_017006892.1 | Intron | XP_016862381.1 | ||||
XM_017006893.1 | Intron | XP_016862382.1 | ||||
XM_017006894.1 | Intron | XP_016862383.1 | ||||
XM_017006895.1 | Intron | XP_016862384.1 | ||||
XM_017006896.1 | Intron | XP_016862385.1 | ||||
XM_017006897.1 | Intron | XP_016862386.1 |
Set Membership: |
HapMap |