Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAACTGTCCTTTGAAAATAGAG[A/G]AATTGGCTGGGTGTGGTGGCTCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607242 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP2A2 - adaptor related protein complex 2 alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242837.1 | Intron | NP_001229766.1 | ||||
NM_012305.3 | Intron | NP_036437.1 | ||||
XM_011519929.1 | Intron | XP_011518231.1 | ||||
XM_011519930.2 | Intron | XP_011518232.1 |
Set Membership: |
HapMap |