Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604036 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNA1 - cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111045.1 | 55 | UTR 5 | NP_001104515.1 | |||
NM_001111046.1 | 55 | Intron | NP_001104516.1 | |||
NM_001111047.1 | 55 | Intron | NP_001104517.1 | |||
NM_003914.3 | 55 | UTR 5 | NP_003905.1 | |||
XM_011535294.2 | 55 | Intron | XP_011533596.1 | |||
XM_011535295.2 | 55 | Intron | XP_011533597.1 | |||
XM_011535296.2 | 55 | Intron | XP_011533598.1 |
Set Membership: |
HapMap |