Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604036 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCNA1 - cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111045.1 | 92 | Intron | NP_001104515.1 | |||
NM_001111046.1 | 92 | Intron | NP_001104516.1 | |||
NM_001111047.1 | 92 | Intron | NP_001104517.1 | |||
NM_003914.3 | 92 | Intron | NP_003905.1 | |||
XM_011535294.2 | 92 | UTR 5 | XP_011533596.1 | |||
XM_011535295.2 | 92 | UTR 5 | XP_011533597.1 | |||
XM_011535296.2 | 92 | Intron | XP_011533598.1 |
Set Membership: |
Validated |