Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614904 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNX7 - sorting nexin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015976.4 | Intron | NP_057060.2 | ||||
NM_152238.3 | Intron | NP_689424.2 | ||||
XM_006710677.2 | Intron | XP_006710740.1 | ||||
XM_006710678.2 | Intron | XP_006710741.1 | ||||
XM_011541564.2 | Intron | XP_011539866.1 | ||||
XM_017001425.1 | Intron | XP_016856914.1 | ||||
XM_017001426.1 | Intron | XP_016856915.1 | ||||
XM_017001427.1 | Intron | XP_016856916.1 | ||||
XM_017001428.1 | Intron | XP_016856917.1 |
Set Membership: |
HapMap |