Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTCTAAGTAGAAAAATTTGCT[C/T]CTGATATAGTAAAAAGTAGGTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605236 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP10D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP10D - ATPase phospholipid transporting 10D (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORIN - corin, serine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278585.1 | Intron | NP_001265514.1 | ||||
NM_001278586.1 | Intron | NP_001265515.1 | ||||
NM_006587.3 | Intron | NP_006578.2 |
Set Membership: |
HapMap |