Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCCATTAGACCTCAGCATTTA[A/T]CCGGAGCGGGGGAGGACGGAGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600576 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GATA4 - GATA binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308093.1 | Intron | NP_001295022.1 | ||||
NM_001308094.1 | Intron | NP_001295023.1 | ||||
NM_002052.4 | Intron | NP_002043.2 | ||||
XM_005272385.4 | Intron | XP_005272442.1 | ||||
XM_005272386.1 | Intron | XP_005272443.1 | ||||
XM_006716248.1 | Intron | XP_006716311.1 | ||||
XM_011543817.2 | Intron | XP_011542119.1 | ||||
XM_011543818.2 | Intron | XP_011542120.1 | ||||
XM_017013312.1 | Intron | XP_016868801.1 |
Set Membership: |
HapMap |