Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACGGACACCCCATCCTACTTC[C/T]GAGGGGGTCCCCAGGTACCTCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612498 MIM: 610982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADSSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADSSL1 - adenylosuccinate synthase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320424.1 | Intron | NP_001307353.1 | ||||
NM_152328.4 | Intron | NP_689541.1 | ||||
NM_199165.2 | Intron | NP_954634.1 | ||||
XM_006720026.3 | Intron | XP_006720089.1 | ||||
XM_011536412.1 | Intron | XP_011534714.1 | ||||
XM_011536414.2 | Intron | XP_011534716.1 |
INF2 - inverted formin, FH2 and WH2 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC388022 - uncharacterized LOC388022 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |