Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAGCCCAAACTCACCCACCCT[C/G]AGTCTAGGTACCCACTTAGCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600068 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT2B7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT2B7 - UDP glucuronosyltransferase family 2 member B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005265702.3 | Intron | XP_005265759.1 | ||||
XM_011532229.2 | Intron | XP_011530531.1 | ||||
XM_011532230.1 | Intron | XP_011530532.1 |