Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC31 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRRC31 - leucine rich repeat containing 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277127.1 | 1253 | UTR 3 | NP_001264056.1 | |||
NM_001277128.1 | 1253 | Silent Mutation | NP_001264057.1 | |||
NM_024727.3 | 1253 | Silent Mutation | NP_079003.2 | |||
XM_011513158.2 | 1253 | Silent Mutation | XP_011511460.1 | |||
XM_011513159.2 | 1253 | Silent Mutation | XP_011511461.1 | |||
XM_011513160.2 | 1253 | Intron | XP_011511462.1 | |||
XM_017007204.1 | 1253 | Silent Mutation | XP_016862693.1 |
LRRIQ4 - leucine rich repeats and IQ motif containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |