Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCATTGGCAATAAATACTGTC[A/C]GGTGTTTTCCATGAAGTGATAGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608662 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ANO5 - anoctamin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142649.1 | Intron | NP_001136121.1 | ||||
NM_213599.2 | Intron | NP_998764.1 | ||||
XM_005252820.3 | Intron | XP_005252877.2 | ||||
XM_005252821.3 | Intron | XP_005252878.2 | ||||
XM_005252822.4 | Intron | XP_005252879.1 | ||||
XM_011519949.2 | Intron | XP_011518251.1 |