Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAACAAACCTGCACTTGTATCTC[C/T]GGACTTAAACTAAAAGTTAACAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO5 - anoctamin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142649.1 | Intron | NP_001136121.1 | ||||
NM_213599.2 | Intron | NP_998764.1 | ||||
XM_005252820.3 | Intron | XP_005252877.2 | ||||
XM_005252821.3 | Intron | XP_005252878.2 | ||||
XM_005252822.4 | Intron | XP_005252879.1 | ||||
XM_011519949.2 | Intron | XP_011518251.1 |