Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATTCAAGGATATGAGAGCAAA[C/T]AAGACCCCATCCTTTTCTCAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611300 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LARP6 - La ribonucleoprotein domain family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC49 - leucine rich repeat containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199017.2 | Intron | NP_001185946.1 | ||||
NM_001199018.2 | Intron | NP_001185947.1 | ||||
NM_001284357.1 | Intron | NP_001271286.1 | ||||
NM_017691.4 | Intron | NP_060161.2 | ||||
XM_005254492.4 | Intron | XP_005254549.1 | ||||
XM_011521715.2 | Intron | XP_011520017.1 | ||||
XM_011521717.2 | Intron | XP_011520019.1 | ||||
XM_011521718.2 | Intron | XP_011520020.1 | ||||
XM_017022357.1 | Intron | XP_016877846.1 | ||||
XM_017022358.1 | Intron | XP_016877847.1 | ||||
XM_017022359.1 | Intron | XP_016877848.1 |