Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605748 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCO1 - beta-carotene oxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017429.2 | Intron | NP_059125.2 | ||||
XM_011523109.1 | Intron | XP_011521411.1 | ||||
XM_017023286.1 | Intron | XP_016878775.1 | ||||
XM_017023287.1 | Intron | XP_016878776.1 | ||||
XM_017023288.1 | Intron | XP_016878777.1 | ||||
XM_017023289.1 | Intron | XP_016878778.1 |
Set Membership: |
HapMap |