Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600407 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RFC5 - replication factor C subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130112.2 | 1519 | Intron | NP_001123584.1 | |||
NM_001206801.1 | 1519 | Intron | NP_001193730.1 | |||
NM_007370.5 | 1519 | Intron | NP_031396.1 | |||
NM_181578.3 | 1519 | Intron | NP_853556.2 | |||
XM_011538643.2 | 1519 | Intron | XP_011536945.1 | |||
XM_011538645.2 | 1519 | Intron | XP_011536947.1 | |||
XM_017019779.1 | 1519 | Intron | XP_016875268.1 |
WSB2 - WD repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278557.1 | 1519 | UTR 3 | NP_001265486.1 | |||
NM_001278558.1 | 1519 | UTR 3 | NP_001265487.1 | |||
NM_018639.4 | 1519 | UTR 3 | NP_061109.1 | |||
XM_017019642.1 | 1519 | UTR 3 | XP_016875131.1 |
Set Membership: |
HapMap Validated |