Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAATGCAGCCATATTTACTTATT[A/G]TCTGTGGTTGTTTTCATGCTATAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123834 MIM: 609514 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF8 - TATA-box binding protein associated factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138572.2 | Intron | NP_612639.2 | ||||
XM_005248849.2 | Intron | XP_005248906.1 | ||||
XM_011514296.1 | Intron | XP_011512598.1 | ||||
XM_017010241.1 | Intron | XP_016865730.1 | ||||
XM_017010242.1 | Intron | XP_016865731.1 | ||||
XM_017010243.1 | Intron | XP_016865732.1 | ||||
XM_017010244.1 | Intron | XP_016865733.1 | ||||
XM_017010245.1 | Intron | XP_016865734.1 |