Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TDRD10 - tudor domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098475.1 | Intron | NP_001091945.1 | ||||
NM_182499.3 | Intron | NP_872305.3 | ||||
XM_006711156.2 | Intron | XP_006711219.1 | ||||
XM_011509152.1 | Intron | XP_011507454.1 | ||||
XM_011509153.1 | Intron | XP_011507455.1 | ||||
XM_011509154.1 | Intron | XP_011507456.1 | ||||
XM_011509155.1 | Intron | XP_011507457.1 | ||||
XM_011509156.1 | Intron | XP_011507458.1 | ||||
XM_011509157.1 | Intron | XP_011507459.1 | ||||
XM_011509158.2 | Intron | XP_011507460.1 | ||||
XM_011509159.2 | Intron | XP_011507461.1 | ||||
XM_011509160.2 | Intron | XP_011507462.1 | ||||
XM_011509161.1 | Intron | XP_011507463.1 | ||||
XM_011509162.2 | Intron | XP_011507464.1 | ||||
XM_017000255.1 | Intron | XP_016855744.1 |